Please login to your online account to display your discounted pricing

AEBSF Hydrochloride, 4-(2-Aminoethyl)-benzenesulfonyl Fluoride HCl, White Powder, Fisher BioReagents


Quantity 1g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26441

 View more versions of this product

Catalog No. BP26441

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Synonym AEBSF HCl
Melting Point 175°C
Physical Form Powder/Solid
Formula Weight 239.7
Quantity 1g
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Color White
Molecular Formula C8H10FNO2S · HCl
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
Purity 97% min.
Assay Percent Range ≥97 %
CAS 30827-99-7
Infrared Spectrum Pass Test
Health Hazard 3 Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:
Identification Pass Test
Packaging Amber Glass
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:DANGER!
Recommended Storage RT, protect from light.

Specifications

Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
CAS 30827-99-7
Physical Form Powder/Solid
Assay Percent Range ≥97 %
Molecular Formula C8H10FNO2S · HCl
Formula Weight 239.7
Synonym AEBSF HCl

  • Irreverisble serine protease inhibitor
  • Inhibits proteases like trypsin, chymotrypsin, kallikrein, plasmin, and thrombin
  • Acts as Sulfonyl fluoride that reacts with hydroxy group of the active site serine residue
  • More soluble and less toxic

Accessories
 

Signal Word:DANGER!

Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:

RT, protect from light.

SDS & Safety Documents