Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Aminoethyl)benzenesulfonamide, 99%

Chemical Identifiers

Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonamide
CAS 35303-76-5
Molecular Formula C8H12N2O2S
Formula Weight 200.26
MDL Number MFCD00010301
UN Number UN3259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2074514

 
alfa aesar™
B2074514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2074522

 
alfa aesar™
B2074522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2074536

 
alfa aesar™
B2074536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.26
Health Hazard 1 Danger
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonamide
Melting Point 150° to 152°C
Assay 99%
Quantity 25g
Molecular Formula C8H12N2O2S
MDL Number MFCD00010301
CAS 35303-76-5
UN Number UN3259
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.