Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2,6-Dimethylphenyl)-3-thiosemicarbazide, 97%

Chemical Identifiers

Chemical Name or Material 4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
CAS 71058-35-0
Molecular Formula C9H13N3S
Formula Weight 195.29
MDL Number MFCD00041281
UN Number UN2811
Beilstein 6768435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1222706

  SDS
alfa aesar™
L1222706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1222703

  SDS
alfa aesar™
L1222703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.29
Chemical Name or Material 4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
Molecular Formula C9H13N3S
Melting Point ≈225°C
MDL Number MFCD00041281
CAS 71058-35-0
Beilstein 6768435
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 97%
UN Number UN2811
Quantity 5g
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.