missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2,6-Dimethylphenyl)-3-thiosemicarbazide, 97%

Manufacturer:  Alfa Aesar™ L1222706

 View more versions of this product

Catalog No. AAL1222706



Specifications

Specifications

4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
5g
C9H13N3S
UN2811
4-2,6-dimethylphenyl-3-thiosemicarbazide, 3-amino-1-2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl hydrazinecarbothioamide, 1-amino-3-2,6-dimethylphenyl thiourea, 2,6-dimethylphenylthiosemicarbazide, 2,6-dimethylphenyl amino hydrazinomethane-1-thione
CC1=C(C(=CC=C1)C)NC(=S)NN
195.284
195.29
~225°C
71058-35-0
MFCD00041281
6768435
YEOAXWBYZFHOIZ-UHFFFAOYSA-N
1-amino-3-(2,6-dimethylphenyl)thiourea
736213
97%

Chemical Identifiers

71058-35-0
195.284
YEOAXWBYZFHOIZ-UHFFFAOYSA-N
736213
CC1=C(C(=CC=C1)C)NC(=S)NN
C9H13N3S
MFCD00041281
4-2,6-dimethylphenyl-3-thiosemicarbazide, 3-amino-1-2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl hydrazinecarbothioamide, 1-amino-3-2,6-dimethylphenyl thiourea, 2,6-dimethylphenylthiosemicarbazide, 2,6-dimethylphenyl amino hydrazinomethane-1-thione
1-amino-3-(2,6-dimethylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents