missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5415603

 View more versions of this product

Catalog No. AAH5415603



Description

Description

Applications
It is an intermediate for the preparation of Elacridar.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline
82925-02-8
MFCD04974539
Soluble in dichloromethane,ethyl acetate and methanol.
COC1=C(C=C2CN(CCC2=C1)CCC3=CC=C(C=C3)N)OC
312.413
312.41
1g
C19H24N2O2
4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine
DGOOLMGPMIHRFY-UHFFFAOYSA-N
4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
10041227
97%

Chemical Identifiers

82925-02-8
312.413
DGOOLMGPMIHRFY-UHFFFAOYSA-N
10041227
COC1=C(C=C2CN(CCC2=C1)CCC3=CC=C(C=C3)N)OC
C19H24N2O2
MFCD04974539
4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine
4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents