missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(2,5-Dimethoxyphenylsulfamoyl)acetanilide, 97%

Chemical Identifiers

CAS 211742-82-4
Molecular Formula C16H18N2O5S
Molecular Weight (g/mol) 350.389
MDL Number MFCD01134327
InChI Key VVYBOPRFGIFZSH-UHFFFAOYSA-N
Synonym n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxy-phenylsulfamoyl-phenyl-acetamide, n-4-2,5-dimethoxyanilino sulfonyl phenyl acetamide, n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxyphenyl amino sulfonyl phenyl acetamide, 4'-2,5-dimethoxyphenylsulfamoyl acetanilide, 4/'-2,5-dimethoxyphenylsulfamoyl acetanilide
PubChem CID 1010065
IUPAC Name N-[4-[(2,5-dimethoxyphenyl)sulfamoyl]phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60663MD View Documents Alfa Aesar™
H60663MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6066303 View Documents Alfa Aesar™
H6066303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Notes
Store in cool dry ventilated place. Ensure proper ventilation.
Specifications

Chemical Identifiers

211742-82-4
350.389
VVYBOPRFGIFZSH-UHFFFAOYSA-N
1010065
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)OC)OC
C16H18N2O5S
MFCD01134327
n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxy-phenylsulfamoyl-phenyl-acetamide, n-4-2,5-dimethoxyanilino sulfonyl phenyl acetamide, n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxyphenyl amino sulfonyl phenyl acetamide, 4'-2,5-dimethoxyphenylsulfamoyl acetanilide, 4/'-2,5-dimethoxyphenylsulfamoyl acetanilide
N-[4-[(2,5-dimethoxyphenyl)sulfamoyl]phenyl]acetamide

Specifications

250mg
C16H18N2O5S
n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxy-phenylsulfamoyl-phenyl-acetamide, n-4-2,5-dimethoxyanilino sulfonyl phenyl acetamide, n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxyphenyl amino sulfonyl phenyl acetamide, 4'-2,5-dimethoxyphenylsulfamoyl acetanilide, 4/'-2,5-dimethoxyphenylsulfamoyl acetanilide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)OC)OC
350.389
350.39
4'-(2,5-Dimethoxyphenylsulfamoyl)acetanilide
211742-82-4
MFCD01134327
VVYBOPRFGIFZSH-UHFFFAOYSA-N
N-[4-[(2,5-dimethoxyphenyl)sulfamoyl]phenyl]acetamide
1010065
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning