missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[2-(4-Morpholinyl)ethyl]-3-thiosemicarbazide, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar L1113003

 View more versions of this product

Catalog No. AAL1113003



Specifications

Specifications

4-[2-(4-Morpholinyl)ethyl]-3-thiosemicarbazide
1g
C7H16N4OS
4733425
OBGPRENQQPWJOC-UHFFFAOYSA-N
1-amino-3-(2-morpholin-4-ylethyl)thiourea
736209
≥98%
164°C to 168°C
77644-45-2
MFCD00041220
4-2-morpholinoethyl-3-thiosemicarbazide, 4-2-morpholinoethyl thiosemicarbazide, 3-amino-1-2-morpholin-4-yl ethyl thiourea, n-2-morpholin-4-yl ethyl hydrazinecarbothioamide, 4-2-4-morpholino ethyl-3-thiosemicarbazide, 1-amino-3-2-morpholin-4-ylethyl thiourea, 1-azanyl-3-2-morpholin-4-ylethyl thiourea, 3-amino-1-2-morpholin-4-ylethyl thiourea, 1-amino-3-2-4-morpholinyl ethyl thiourea, 4-2-4-morpholinyl ethyl-3-thiosemicarbazide
C1COCCN1CCNC(=S)NN
204.292
204.3

Chemical Identifiers

77644-45-2
204.292
OBGPRENQQPWJOC-UHFFFAOYSA-N
736209
C1COCCN1CCNC(=S)NN
C7H16N4OS
MFCD00041220
4-2-morpholinoethyl-3-thiosemicarbazide, 4-2-morpholinoethyl thiosemicarbazide, 3-amino-1-2-morpholin-4-yl ethyl thiourea, n-2-morpholin-4-yl ethyl hydrazinecarbothioamide, 4-2-4-morpholino ethyl-3-thiosemicarbazide, 1-amino-3-2-morpholin-4-ylethyl thiourea, 1-azanyl-3-2-morpholin-4-ylethyl thiourea, 3-amino-1-2-morpholin-4-ylethyl thiourea, 1-amino-3-2-4-morpholinyl ethyl thiourea, 4-2-4-morpholinyl ethyl-3-thiosemicarbazide
1-amino-3-(2-morpholin-4-ylethyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents