Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2,4-Dimethylphenyl)-3-thiosemicarbazide, 98%

Chemical Identifiers

Chemical Name or Material 4-(2,4-Dimethylphenyl)-3-thiosemicarbazide
CAS 66298-09-7
Molecular Formula C9H13N3S
Formula Weight 195.29
MDL Number MFCD00041279
UN Number UN2811
Beilstein 2644678
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1033803

 
alfa aesar™
L1033803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1033806

 
alfa aesar™
L1033806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.29
Molecular Formula C9H13N3S
Chemical Name or Material 4-(2,4-Dimethylphenyl)-3-thiosemicarbazide
Assay 98%
MDL Number MFCD00041279
CAS 66298-09-7
UN Number UN2811
Beilstein 2644678
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Melting Point 150° to 152°C
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.