missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2,4-Dimethylphenyl)-3-thiosemicarbazide, 98%

Manufacturer:  Alfa Aesar™ L1033803

 View more versions of this product

Catalog No. AAL1033803



Specifications

Specifications

4-(2,4-Dimethylphenyl)-3-thiosemicarbazide
1g
C9H13N3S
UN2811
4-2,4-dimethylphenyl-3-thiosemicarbazide, 3-amino-1-2,4-dimethylphenyl thiourea, n-2,4-dimethylphenyl hydrazinecarbothioamide, 1-amino-3-2,4-dimethylphenyl thiourea, 2,4-dimethylphenyl amino hydrazinomethane-1-thione, 4-2,4-dimethylphenyl thiosemicarbazide, 1-azanyl-3-2,4-dimethylphenyl thiourea
CC1=CC(=C(C=C1)NC(=S)NN)C
195.284
195.29
150°C to 152°C
66298-09-7
MFCD00041279
2644678
YTRJQDFRZLKDHZ-UHFFFAOYSA-N
1-amino-3-(2,4-dimethylphenyl)thiourea
737131
98%

Chemical Identifiers

66298-09-7
195.284
YTRJQDFRZLKDHZ-UHFFFAOYSA-N
737131
CC1=CC(=C(C=C1)NC(=S)NN)C
C9H13N3S
MFCD00041279
4-2,4-dimethylphenyl-3-thiosemicarbazide, 3-amino-1-2,4-dimethylphenyl thiourea, n-2,4-dimethylphenyl hydrazinecarbothioamide, 1-amino-3-2,4-dimethylphenyl thiourea, 2,4-dimethylphenyl amino hydrazinomethane-1-thione, 4-2,4-dimethylphenyl thiosemicarbazide, 1-azanyl-3-2,4-dimethylphenyl thiourea
1-amino-3-(2,4-dimethylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents