Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2,4-Dichlorophenyl)-3-thiosemicarbazide, 98+%

Chemical Identifiers

Chemical Name or Material 4-(2,4-Dichlorophenyl)-3-thiosemicarbazide
CAS 13124-11-3
Molecular Formula C7H7Cl2N3S
Formula Weight 236.12
MDL Number MFCD00041272
UN Number UN2811
Beilstein 651150
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1259903

  SDS
alfa aesar™
L1259903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1259906

  SDS
alfa aesar™
L1259906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.12
MDL Number MFCD00041272
CAS 13124-11-3
Chemical Name or Material 4-(2,4-Dichlorophenyl)-3-thiosemicarbazide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay ≥98%
Molecular Formula C7H7Cl2N3S
Melting Point 167° to 170°C
Quantity 1g
UN Number UN2811
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Beilstein 651150
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.