missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2,3-Dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 54557-81-2
Molecular Formula C12H12O5
Molecular Weight (g/mol) 236.223
MDL Number MFCD00168683
InChI Key LMDXEMFSAHAGGP-UHFFFAOYSA-N
Synonym 4-2,3-dihydro-1,4-benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydro-benzo 1,4 dioxin-6-yl-4-oxo-butyric acid, 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-4-oxobutanoic acid, 4-3,4-ethylenedioxy phenyl-4-oxobutyric acid, 4-2,3-dihydro-1,4,benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydrobenzo b 1,4 dioxin-7-yl-4-oxobutanoic acid, 4-oxo-4-2,3-dihydro-1,4-benzodioxin-6-yl butyric acid, 4-2h,3h-benzo 3,4-e 1,4-dioxin-6-yl-4-oxobutanoic acid, maybridge1_001974, regid_for_cid_2758835
PubChem CID 2758835
IUPAC Name 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid
SMILES C1COC2=C(O1)C=CC(=C2)C(=O)CCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB12838DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
BTB12838DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB12838EA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
BTB12838EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

54557-81-2
236.223
LMDXEMFSAHAGGP-UHFFFAOYSA-N
2758835
C1COC2=C(O1)C=CC(=C2)C(=O)CCC(=O)O
C12H12O5
MFCD00168683
4-2,3-dihydro-1,4-benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydro-benzo 1,4 dioxin-6-yl-4-oxo-butyric acid, 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-4-oxobutanoic acid, 4-3,4-ethylenedioxy phenyl-4-oxobutyric acid, 4-2,3-dihydro-1,4,benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydrobenzo b 1,4 dioxin-7-yl-4-oxobutanoic acid, 4-oxo-4-2,3-dihydro-1,4-benzodioxin-6-yl butyric acid, 4-2h,3h-benzo 3,4-e 1,4-dioxin-6-yl-4-oxobutanoic acid, maybridge1_001974, regid_for_cid_2758835
4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid

Specifications

54557-81-2
100.0
Amber Glass Bottle
MFCD00168683
4-2,3-dihydro-1,4-benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydro-benzo 1,4 dioxin-6-yl-4-oxo-butyric acid, 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-4-oxobutanoic acid, 4-3,4-ethylenedioxy phenyl-4-oxobutyric acid, 4-2,3-dihydro-1,4,benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydrobenzo b 1,4 dioxin-7-yl-4-oxobutanoic acid, 4-oxo-4-2,3-dihydro-1,4-benzodioxin-6-yl butyric acid, 4-2h,3h-benzo 3,4-e 1,4-dioxin-6-yl-4-oxobutanoic acid, maybridge1_001974, regid_for_cid_2758835
C1COC2=C(O1)C=CC(=C2)C(=O)CCC(=O)O
236.223
236.22
4-(2, 3-Dihydro-1, 4-benzodioxin-6-yl)-4-oxobutanoic acid
96.5
97%
C12H12O5
1g
LMDXEMFSAHAGGP-UHFFFAOYSA-N
4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid
2758835
97%
SDS
missing translation for 'documents'

missing translation for 'documents'