missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H53245MD

 View more versions of this product

Catalog No. AAH53245MD



Specifications

Specifications

4-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride
250mg
C8H9BClN5O3
4-1h-tetrazol-5-yl carbamoyl phenyl boronic acid hydrochloride, 4-1h-tetrazol-5-ylcarbamoyl benzeneboronic acid hydrochloride, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid, hcl, 4-1h-tetrazol-5-yl-carbamoyl benzeneboronic acid, hcl, 4-1h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hcl, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hydrochloride, 4-1h-tetrazol-5-yl carbamoyl phenyl boronicacidhydrochloride, 4-1h-tetrazol-5-ylaminocarbonyl phenylboronic acid hydrochloride, 4-2h-tetrazol-5-yl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
B(C1=CC=C(C=C1)C(=O)NC2=NNN=N2)(O)O.Cl
269.452
269.45
>300°C
850568-31-9
MFCD06659882
AZODLVUFJXZNTC-UHFFFAOYSA-N
[4-(2H-tetrazol-5-ylcarbamoyl)phenyl]boronic acid;hydrochloride
44119407
97%

Chemical Identifiers

850568-31-9
269.452
AZODLVUFJXZNTC-UHFFFAOYSA-N
44119407
B(C1=CC=C(C=C1)C(=O)NC2=NNN=N2)(O)O.Cl
C8H9BClN5O3
MFCD06659882
4-1h-tetrazol-5-yl carbamoyl phenyl boronic acid hydrochloride, 4-1h-tetrazol-5-ylcarbamoyl benzeneboronic acid hydrochloride, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid, hcl, 4-1h-tetrazol-5-yl-carbamoyl benzeneboronic acid, hcl, 4-1h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hcl, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hydrochloride, 4-1h-tetrazol-5-yl carbamoyl phenyl boronicacidhydrochloride, 4-1h-tetrazol-5-ylaminocarbonyl phenylboronic acid hydrochloride, 4-2h-tetrazol-5-yl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
[4-(2H-tetrazol-5-ylcarbamoyl)phenyl]boronic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents