missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1H-Tetrazol-5-yl)phenol, 97%

$100.84 - $759.93

Chemical Identifiers

CAS 51517-88-5
Molecular Formula C7H6N4O
Formula Weight 162.16
MDL Number MFCD05662709
Synonym 5-(4-Hydroxyphenyl)-1H-tetrazole
Chemical Name or Material 4-(1H-Tetrazol-5-yl)phenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2691806 SDS Alfa Aesar™
H2691806
5g Each for $759.93
AAH26918MD SDS Alfa Aesar™
H26918MD
250mg Each for $100.84
AAH2691803 SDS Alfa Aesar™
H2691803
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

51517-88-5
162.16
5-(4-Hydroxyphenyl)-1H-tetrazole
C7H6N4O
MFCD05662709
4-(1H-Tetrazol-5-yl)phenol

Specifications

≈240°C
51517-88-5
162.16
5-(4-Hydroxyphenyl)-1H-tetrazole
97%
5g
C7H6N4O
MFCD05662709
4-(1H-Tetrazol-5-yl)phenol

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning