Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1H-Tetrazol-5-yl)benzyl alcohol, 97%

$115.12 - $115.12

Chemical Identifiers

Chemical Name or Material 4-(1H-Tetrazol-5-yl)benzyl alcohol
CAS 501126-02-9
Molecular Formula C8H8N4O
Formula Weight 176.18
MDL Number MFCD09909570
Synonym 5-(4-Hydroxymethylphenyl)-1H-tetrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26964MD

  SDS
alfa aesar™
H26964MD
250mg Each for $115.12
Description & Specifications

Specifications

Formula Weight 176.18
Chemical Name or Material 4-(1H-Tetrazol-5-yl)benzyl alcohol
Assay 97%
Molecular Formula C8H8N4O
MDL Number MFCD09909570
CAS 501126-02-9
Synonym 5-(4-Hydroxymethylphenyl)-1H-tetrazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.