Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1H-Tetrazol-5-yl)aniline, 97%

$109.24 - $825.70

Chemical Identifiers

Chemical Name or Material 4-(1H-Tetrazol-5-yl)aniline
CAS 46047-18-1
Molecular Formula C7H7N5
Formula Weight 161.17
MDL Number MFCD01219548
Synonym 5-(4-Aminophenyl)-1H-tetrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2731106

  SDS
alfa aesar™
H2731106
5g Each for $825.70

AAH2731103

  SDS
alfa aesar™
H2731103
1g Each for $274.23

AAH27311MD

  SDS
alfa aesar™
H27311MD
250mg Each for $109.24
Description & Specifications

Specifications

Formula Weight 161.17
Chemical Name or Material 4-(1H-Tetrazol-5-yl)aniline
Assay 97%
Molecular Formula C7H7N5
MDL Number MFCD01219548
CAS 46047-18-1
Synonym 5-(4-Aminophenyl)-1H-tetrazole
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈265°C
Quantity 5g

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.