Please login to your online account to display your discounted pricing

4-(1H-Pyrrol-1-yl)phenol, 97%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC372720010

 View more versions of this product

Catalog No. AC372720010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 159.19
Quantity 1g
Chemical Name or Material 4-(1H-Pyrrol-1-yl)phenol
Assay Percent Range 97%
Molecular Formula C10H9NO
CAS 23351-09-9
Packaging Glass bottle

Specifications

Chemical Name or Material 4-(1H-Pyrrol-1-yl)phenol
Assay Percent Range 97%
Molecular Formula C10H9NO
Formula Weight 159.19