Please login to your online account to display your discounted pricing

4-(1H-Pyrrol-1-yl)phenol, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-(1H-Pyrrol-1-yl)phenol
Assay Percent Range 97%
Molecular Formula C10H9NO
Formula Weight 159.19
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC372720010

  SDS
thermo fisher scientific
AC372720010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.19
Quantity 1g
Packaging Glass bottle
CAS 23351-09-9
Chemical Name or Material 4-(1H-Pyrrol-1-yl)phenol
Assay Percent Range 97%
Molecular Formula C10H9NO