Please login to your online account to display your discounted pricing

4-(1H-Pyrrol-1-yl)benzylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(1H-pyrrol-1-yl)benzylamine
CAS 465514-27-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H12N2
Formula Weight 172.23
MDL Number MFCD03659728
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC25513CB

  SDS
fisher scientific company
CC25513CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC25513DA

  SDS
fisher scientific company
CC25513DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.23
Quantity 250mg
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
MDL Number MFCD03659728
CAS Min % 96.5
Chemical Name or Material 4-(1H-pyrrol-1-yl)benzylamine
Assay Percent Range 97%
Molecular Formula C11H12N2
CAS 465514-27-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.