Please login to your online account to display your discounted pricing

Alfa Aesar™ 4(1H)-Pyrimidinone, 98%

Chemical Identifiers

Chemical Name or Material 4(1H)-Pyrimidinone
CAS 4562-27-0
Formula Weight 96.09
MDL Number MFCD00006664
Beilstein 741972
Synonym 4-Pyrimidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6469003

  SDS
alfa aesar™
H6469003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6469006

  SDS
alfa aesar™
H6469006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 163° to 169°C (lit.)
Formula Weight 96.09
Chemical Name or Material 4(1H)-Pyrimidinone
Assay 98%
Synonym 4-Pyrimidinol
Physical Form Solid
MDL Number MFCD00006664
Beilstein 741972
RTECS Number UW7350100
CAS 4562-27-0
TSCA TSCA
Storage Note 1 2° to 8°C
Quantity 1g
Safety and Handling

2° to 8°C