Please login to your online account to display your discounted pricing

[4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol 97+%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC56009DA

 View more versions of this product

Catalog No. CC56009DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 188.23
Quantity 1g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material [4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
Packaging Amber Glass Bottle
Molecular Formula C11H12N2O
CAS 160388-55-6

Specifications

Chemical Name or Material [4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H12N2O
Formula Weight 188.23