Please login to your online account to display your discounted pricing

4-(1H-Pyrazol-1-ylmethyl)benzoic acid, ≥97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC56001DA

 View more versions of this product

Catalog No. CC56001DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.21
Quantity 1g
Assay 97%
CAS Max % 100.0
CAS 160388-53-4
Molecular Formula C11H10N2O2
MDL Number MFCD07186451
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzoic acid

Specifications

Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzoic acid
CAS 160388-53-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O2
Formula Weight 202.21
MDL Number MFCD07186451
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents