Please login to your online account to display your discounted pricing

4-(1H-pyrazol-1-ylmethyl)benzaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O
Formula Weight 186.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC56004DA

  SDS
fisher scientific company
CC56004DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC56004DE

 
fisher scientific company
CC56004DE
Amber glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC56004CB

  SDS
fisher scientific company
CC56004CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.21
Quantity 1g
Assay 97%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Assay Percent Range 97%
Molecular Formula C11H10N2O
CAS 887922-90-9
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning