missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[4-(1H-Pyrazol-1-yl)phenyl]methanol, 97%, Maybridge

Chemical Identifiers

CAS 143426-49-7
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD02682057
InChI Key MMGMLIHSHFKRDK-UHFFFAOYSA-N
Synonym 4-1-pyrazolyl benzylalcohol
PubChem CID 2776480
IUPAC Name (4-pyrazol-1-ylphenyl)methanol
SMILES C1=CN(N=C1)C2=CC=C(C=C2)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC18409DA SDS View Documents Fisher Scientific Company
CC18409DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

143426-49-7
174.203
MMGMLIHSHFKRDK-UHFFFAOYSA-N
2776480
C1=CN(N=C1)C2=CC=C(C=C2)CO
C10H10N2O
MFCD02682057
4-1-pyrazolyl benzylalcohol
(4-pyrazol-1-ylphenyl)methanol

Specifications

Amber Glass Bottle
143426-49-7
100.0
C10H10N2O
4-1-pyrazolyl benzylalcohol
C1=CN(N=C1)C2=CC=C(C=C2)CO
174.203
174.2
[4-(1H-Pyrazol-1-yl)phenyl]methanol
1g
96.5
97%
MFCD02682057
MMGMLIHSHFKRDK-UHFFFAOYSA-N
(4-pyrazol-1-ylphenyl)methanol
2776480
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning