Please login to your online account to display your discounted pricing

4-(1H-Pyrazol-1-yl)benzaldehyde, ≥95%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC18404DA

 View more versions of this product

Catalog No. CC18404DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.19
Quantity 1g
Assay 95%
CAS Max % 100.0
CAS 99662-34-7
Assay Percent Range 95%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzaldehyde
MDL Number MFCD02681938
CAS Min % 94.5
Packaging Amber glass bottle
Molecular Formula C10H8N2O

Specifications

Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzaldehyde
CAS 99662-34-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H8N2O
Formula Weight 172.19
MDL Number MFCD02681938