Please login to your online account to display your discounted pricing

4-(1H-Pyrazol-1-yl)benzaldehyde, ≥95%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  CC18404EB

 View more versions of this product

Catalog No. CC18404EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.19
Quantity 25g
Assay 95%
CAS Max % 100.0
Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzaldehyde
CAS 99662-34-7
Assay Percent Range 95%
Molecular Formula C10H8N2O
CAS Min % 94.5
MDL Number MFCD02681938

Specifications

Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzaldehyde
CAS 99662-34-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H8N2O
Formula Weight 172.19
MDL Number MFCD02681938