missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1h-imidazol-1-ylmethyl)benzoic acid hydrochloride, Maybridge

$67.08 - $67.08

Chemical Identifiers

CAS 135611-32-4
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H10N2O2·ClH
Formula Weight 238.67
MDL Number MFCD08271922
Chemical Name or Material 4-(1H-imidazol-1-ylmethyl)benzoic acid hydrochloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC43351CB SDS Fisher Scientific Company
CC43351CB
Amber Glass Bottle 250mg Each for $67.08
CC43351DA SDS Fisher Scientific Company
CC43351DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

135611-32-4
100.0
C11H10N2O2·ClH
MFCD08271922
95%
94.5
95%
238.67
4-(1H-imidazol-1-ylmethyl)benzoic acid hydrochloride

Specifications

238.67
100.0
Amber Glass Bottle
MFCD08271922
94.5
95%
250mg
C11H10N2O2·ClH
95%
135611-32-4
4-(1H-imidazol-1-ylmethyl)benzoic acid hydrochloride

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.