Please login to your online account to display your discounted pricing

4-(1h-Imidazol-1-ylmethyl)benzaldehyde, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  CC43304DA

 View more versions of this product

Catalog No. CC43304DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 186.21
Quantity 1g
Assay 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)benzaldehyde
Assay Percent Range 97%
Molecular Formula C11H10N2O
CAS 102432-03-1

Specifications

Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)benzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O
Formula Weight 186.21