missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[4-(1H-Imidazol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC18509EA

 View more versions of this product

Catalog No. CC18509EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

[4-(1H-Imidazol-1-yl)phenyl]methanol
96.5
97%
MFCD02682059
4-1h-imidazol-1-yl phenyl methanol, 4-imidazol-1-yl-phenyl methanol, 4-1-imidazolyl phenyl methanol, 4-imidazol-1-ylphenyl methanol, 4-1-imidazolyl benzyl alcohol, 4-imidazol-1-yl phenyl methanol, benzenemethanol, 4-1h-imidazol-1-yl, acmc-209qb5, 4-imidazolylphenyl methan-1-ol
C1=CC(=CC=C1CO)N2C=CN=C2
174.203
174.2
86718-08-3
100.0
C10H10N2O
10g
SRQXVPAXMMIUFH-UHFFFAOYSA-N
(4-imidazol-1-ylphenyl)methanol
2776486
97%

Chemical Identifiers

86718-08-3
174.203
SRQXVPAXMMIUFH-UHFFFAOYSA-N
2776486
C1=CC(=CC=C1CO)N2C=CN=C2
C10H10N2O
MFCD02682059
4-1h-imidazol-1-yl phenyl methanol, 4-imidazol-1-yl-phenyl methanol, 4-1-imidazolyl phenyl methanol, 4-imidazol-1-ylphenyl methanol, 4-1-imidazolyl benzyl alcohol, 4-imidazol-1-yl phenyl methanol, benzenemethanol, 4-1h-imidazol-1-yl, acmc-209qb5, 4-imidazolylphenyl methan-1-ol
(4-imidazol-1-ylphenyl)methanol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions