missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1H-Imidazol-1-yl)benzoic acid, ≥90%, Maybridge

Chemical Identifiers

CAS 17616-04-5
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00586645
InChI Key LFIDZIWWYNTQOQ-UHFFFAOYSA-N
Synonym 4-1-imidazolyl benzoic acid
PubChem CID 736529
IUPAC Name 4-imidazol-1-ylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=CN=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC18501DA SDS View Documents Fisher Scientific Company
CC18501DA
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18501EA SDS View Documents Fisher Scientific Company
CC18501EA
10g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

17616-04-5
188.186
LFIDZIWWYNTQOQ-UHFFFAOYSA-N
736529
C1=CC(=CC=C1C(=O)O)N2C=CN=C2
C10H8N2O2
MFCD00586645
4-1-imidazolyl benzoic acid
4-imidazol-1-ylbenzoic acid

Specifications

Amber glass bottle
17616-04-5
100.0
C10H8N2O2
4-1-imidazolyl benzoic acid
C1=CC(=CC=C1C(=O)O)N2C=CN=C2
188.186
188.19
4-(1H-imidazol-1-yl)benzoic acid
1g
89.5
90%
MFCD00586645
LFIDZIWWYNTQOQ-UHFFFAOYSA-N
4-imidazol-1-ylbenzoic acid
736529
90%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.