Please login to your online account to display your discounted pricing

4-(1H-Imidazol-1-yl)benzaldehyde, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC18504CB

 View more versions of this product

Catalog No. CC18504CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.19
Quantity 250mg
Assay 97%
CAS Max % 100.0
Molecular Formula C10H8N2O
Assay Percent Range 97%
CAS 10040-98-9
CAS Min % 96.5
MDL Number MFCD00209977
Packaging Amber Glass Bottle
Chemical Name or Material 4-(1H-Imidazol-1-yl)benzaldehyde

Specifications

Chemical Name or Material 4-(1H-Imidazol-1-yl)benzaldehyde
CAS 10040-98-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O
Formula Weight 172.19
MDL Number MFCD00209977