Please login to your online account to display your discounted pricing

4-(1H-Imidazol-1-yl)benzaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(1H-Imidazol-1-yl)benzaldehyde
CAS 10040-98-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O
Formula Weight 172.19
MDL Number MFCD00209977
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC18504CB

  SDS
fisher scientific company
CC18504CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.19
Quantity 250mg
Assay 97%
CAS Max % 100.0
Molecular Formula C10H8N2O
Assay Percent Range 97%
CAS 10040-98-9
CAS Min % 96.5
MDL Number MFCD00209977
Packaging Amber Glass Bottle
Chemical Name or Material 4-(1H-Imidazol-1-yl)benzaldehyde