Please login to your online account to display your discounted pricing

4-(1H-Benzimidazol-1-ylmethyl)benzoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(1H-Benzimidazol-1-ylmethyl)benzoic acid
CAS 139742-50-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H12N2O2
Formula Weight 252.27
MDL Number MFCD02180372
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

KM10745EA

 
fisher scientific company
KM10745EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.27
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-(1H-Benzimidazol-1-ylmethyl)benzoic acid
Assay Percent Range 97%
Molecular Formula C15H12N2O2
MDL Number MFCD02180372
CAS 139742-50-0