missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1H-1,2,4-triazol-1-yl)phenol, Maybridge™

Chemical Identifiers

CAS 68337-15-5
Molecular Formula C8H7N3O
Formula Weight 161.16308
MDL Number MFCD00003096
Chemical Name or Material 4-(1H-1, 2, 4-triazol-1-yl)phenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC18635EA SDS Fisher Scientific Company
AC18635EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC18635EE SDS Fisher Scientific Company
AC18635EE
Amber Glass Bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

68337-15-5
161.16308
4-(1H-1, 2, 4-triazol-1-yl)phenol
C8H7N3O
MFCD00003096

Specifications

68337-15-5
C8H7N3O
MFCD00003096
4-(1H-1, 2, 4-triazol-1-yl)phenol
Amber Glass Bottle
161.16308
10g