Please login to your online account to display your discounted pricing

4-(1H-1,2,4-triazol-1-yl)phenol, Maybridge™


Quantity 50g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AC18635EE

 View more versions of this product

Catalog No. AC18635EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.16308
Quantity 50g
Packaging Amber Glass Bottle
Molecular Formula C8H7N3O
Chemical Name or Material 4-(1H-1, 2, 4-triazol-1-yl)phenol
CAS 68337-15-5
MDL Number MFCD00003096

Specifications

Chemical Name or Material 4-(1H-1, 2, 4-triazol-1-yl)phenol
CAS 68337-15-5
Molecular Formula C8H7N3O
Formula Weight 161.16308
MDL Number MFCD00003096