Please login to your online account to display your discounted pricing

4-(1H-1,2,4-Triazol-1-yl)benzaldehyde, 95+%

Chemical Identifiers

Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3O
Formula Weight 173.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC18604DA

  SDS
fisher scientific company
CC18604DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC18604DE

  SDS
fisher scientific company
CC18604DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.17
Quantity 1g
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3O
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Packaging Amber Glass Bottle
CAS 27996-86-7
Health Hazard 1 Warning
Safety and Handling

Warning