Please login to your online account to display your discounted pricing

4-(1H-1,2,4-Triazol-1-yl)benzaldehyde, 95+%


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC18604DE

 View more versions of this product

Catalog No. CC18604DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.17
Quantity 5g
Assay 95%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Packaging Amber Glass Bottle
Molecular Formula C9H7N3O
Health Hazard 1 Warning
Assay Percent Range 95%
CAS 27996-86-7

Specifications

Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3O
Formula Weight 173.17
Accessories

Warning