Please login to your online account to display your discounted pricing

4-(1H-1,2,4-Triazol-1-yl)benzaldehyde, 95+%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC18604DA

 View more versions of this product

Catalog No. CC18604DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.17
Quantity 1g
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3O
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Packaging Amber Glass Bottle
CAS 27996-86-7
Health Hazard 1 Warning

Specifications

Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3O
Formula Weight 173.17
Accessories

Warning