Learn More
4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid, ≥90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals MO00953DA
Specifications
4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid | |
97.0 | |
C17H23NO5 | |
1g | |
LUVXDMFUUZGFST-UHFFFAOYSA-N | |
4-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxybenzoic acid | |
2794649 | |
97% |
162046-56-2 | |
97% | |
MFCD06658985 | |
4-1-tert-butoxycarbonyl piperidin-4-yloxy benzoic acid, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy benzoic acid, 4-1-boc-4-piperidyloxy benzoic acid, 4-1-tert-butoxycarbonyl-4-piperidinyl oxy benzoic acid, 4-1-boc piperidin-4-yloxy benzoic acid, 4-4-carboxyphenoxy piperidine, n-boc protected, 4-1-t-butoxycarbonyl-4-piperidyloxy benzoic acid, 4 1-t-butoxycarbonyl 4-piperadinyl oxybenzoic acid | |
CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=C(C=C2)C(=O)O | |
321.373 | |
321.37 |
Chemical Identifiers
162046-56-2 | |
321.373 | |
LUVXDMFUUZGFST-UHFFFAOYSA-N | |
2794649 | |
CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=C(C=C2)C(=O)O |
C17H23NO5 | |
MFCD06658985 | |
4-1-tert-butoxycarbonyl piperidin-4-yloxy benzoic acid, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy benzoic acid, 4-1-boc-4-piperidyloxy benzoic acid, 4-1-tert-butoxycarbonyl-4-piperidinyl oxy benzoic acid, 4-1-boc piperidin-4-yloxy benzoic acid, 4-4-carboxyphenoxy piperidine, n-boc protected, 4-1-t-butoxycarbonyl-4-piperidyloxy benzoic acid, 4 1-t-butoxycarbonyl 4-piperadinyl oxybenzoic acid | |
4-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxybenzoic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning