Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Pyrrolyl)benzoic acid, 99%

$137.73 - $137.73

Chemical Identifiers

Chemical Name or Material 4-(1-Pyrrolyl)benzoic acid
CAS 22106-33-8
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00082569
Synonym 1-(4-Carboxyphenyl)pyrrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107303

  SDS
alfa aesar™
H5107303
1g Each for $137.73
Description & Specifications

Specifications

Formula Weight 187.2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-(1-Pyrrolyl)benzoic acid
Health Hazard 1 Warning
Molecular Formula C11H9NO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
MDL Number MFCD00082569
CAS 22106-33-8
Synonym 1-(4-Carboxyphenyl)pyrrole
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.