Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester

$95.28 - $364.81

Chemical Identifiers

Chemical Name or Material 4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester
CAS 884507-39-5
Molecular Formula C17H26BNO2
Formula Weight 287.2
MDL Number MFCD08690299
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5111503

 
alfa aesar™
H5111503
1g Each for $95.28

AAH5111506

 
alfa aesar™
H5111506
5g Each for $364.81
Description & Specifications

Specifications

Formula Weight 287.2
Chemical Name or Material 4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester
Molecular Formula C17H26BNO2
MDL Number MFCD08690299
CAS 884507-39-5
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
Quantity 1g