missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1-Pyrrolidinyl)piperidine, TCI America™

Manufacturer:  TCI America P193225G

 View more versions of this product

Catalog No. P193225G



Specifications

Specifications

4-(1-Pyrrolidinyl)piperidine
60°C
5004-07-9
MFCD00038011
STWODXDTKGTVCJ-UHFFFAOYSA-N
4-pyrrolidin-1-ylpiperidine
78703
Crystal-Powder at 20°C
White-Yellow
25g
C9H18N2
4-1-pyrrolidinyl piperidine, 4-pyrrolidin-1-yl piperidine, 4-pyrrolidin-1-yl-piperidine, 4-pyrrolidinopiperidine, 4-1-pyrrolidinyl-piperidine, piperidine, 4-1-pyrrolidinyl, 4-pyrrolidinylpiperidine, 4-1-pyrrolidinyl piperidine, 4-pyrrolizinopiperidine, 4-pyrrolidino-piperidine
C1CCN(C1)C2CCNCC2
154.26
154.26
≥98.0% (GC,T)

Chemical Identifiers

5004-07-9
154.26
STWODXDTKGTVCJ-UHFFFAOYSA-N
78703
C1CCN(C1)C2CCNCC2
C9H18N2
MFCD00038011
4-1-pyrrolidinyl piperidine, 4-pyrrolidin-1-yl piperidine, 4-pyrrolidin-1-yl-piperidine, 4-pyrrolidinopiperidine, 4-1-pyrrolidinyl-piperidine, piperidine, 4-1-pyrrolidinyl, 4-pyrrolidinylpiperidine, 4-1-pyrrolidinyl piperidine, 4-pyrrolizinopiperidine, 4-pyrrolidino-piperidine
4-pyrrolidin-1-ylpiperidine
Safety and Handling

Safety and Handling

TSCA : No

Documents