Please login to your online account to display your discounted pricing

4-(1-Pyrrolidinyl)benzoic acid, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC30101DA

 View more versions of this product

Catalog No. CC30101DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 191.23
Quantity 1g
Assay 97%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzoic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H13NO2
CAS 22090-27-3
Health Hazard 1 Warning

Specifications

Chemical Name or Material 4-(1-Pyrrolidinyl)benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H13NO2
Formula Weight 191.23
Accessories

Warning