missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester, 97%

CAS: 852227-90-8 | C16H24BNO2 | 273.18 g/mol

$134.33 - $457.22

Chemical Identifiers

CAS 852227-90-8
Molecular Formula C16H24BNO2
Molecular Weight (g/mol) 273.18
MDL Number MFCD08060504
InChI Key DWJNNJSONWFVBT-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
PubChem CID 18525715
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59911MD
View Documents
Thermo Scientific Chemicals
H59911MD
250 mg
Each for $134.33
Only null left
 
AAH5991103
View Documents
Thermo Scientific Chemicals
H5991103
1 g
Each for $457.22
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

852227-90-8
273.18
DWJNNJSONWFVBT-UHFFFAOYSA-N
18525715
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1
C16H24BNO2
MFCD08060504
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine

Specifications

852227-90-8
MFCD08060504
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
DWJNNJSONWFVBT-UHFFFAOYSA-N
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
18525715
97%
C16H24BNO2
250 mg
Slightly soluble in water.
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1
273.18
273.18
4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only