missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Pyrrolidinyl)benzaldehyde, 98+%

$82.65 - $282.20

Chemical Identifiers

CAS 51980-54-2
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00015469
InChI Key DATXHLPRESKQJK-UHFFFAOYSA-N
Synonym 4-1-pyrrolidinyl benzaldehyde, 4-pyrrolidin-1-yl benzaldehyde, 4-1-pyrrolidino benzaldehyde, 1-4-formylphenyl pyrrolidine, benzaldehyde, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-benzaldehyde, 4-pyrrolidinobenzaldehyde, 4-pyrrolidinylbenzaldehyde, 4-1-pyrrolidino-benzaldehyde, 4-pyrrolizinobenzaldehyde
PubChem CID 104037
IUPAC Name 4-pyrrolidin-1-ylbenzaldehyde
SMILES C1CCN(C1)C2=CC=C(C=C2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0096606 View Documents Alfa Aesar™
L0096606
5g Each for $82.65
 
AAL0096614 View Documents Alfa Aesar™
L0096614
25g Each for $282.20
 
Specifications

Chemical Identifiers

51980-54-2
175.231
DATXHLPRESKQJK-UHFFFAOYSA-N
104037
C1CCN(C1)C2=CC=C(C=C2)C=O
C11H13NO
MFCD00015469
4-1-pyrrolidinyl benzaldehyde, 4-pyrrolidin-1-yl benzaldehyde, 4-1-pyrrolidino benzaldehyde, 1-4-formylphenyl pyrrolidine, benzaldehyde, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-benzaldehyde, 4-pyrrolidinobenzaldehyde, 4-pyrrolidinylbenzaldehyde, 4-1-pyrrolidino-benzaldehyde, 4-pyrrolizinobenzaldehyde
4-pyrrolidin-1-ylbenzaldehyde

Specifications

82°C to 85°C
51980-54-2
MFCD00015469
4-1-pyrrolidinyl benzaldehyde, 4-pyrrolidin-1-yl benzaldehyde, 4-1-pyrrolidino benzaldehyde, 1-4-formylphenyl pyrrolidine, benzaldehyde, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-benzaldehyde, 4-pyrrolidinobenzaldehyde, 4-pyrrolidinylbenzaldehyde, 4-1-pyrrolidino-benzaldehyde, 4-pyrrolizinobenzaldehyde
C1CCN(C1)C2=CC=C(C=C2)C=O
175.231
175.23
4-(1-Pyrrolidinyl)benzaldehyde
5g
C11H13NO
135279
DATXHLPRESKQJK-UHFFFAOYSA-N
4-pyrrolidin-1-ylbenzaldehyde
104037
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C