missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(1-Pyrrolidinyl)acetophenone, 98+%

$84.22 - $294.70

Chemical Identifiers

CAS 21557-09-5
Molecular Formula C12H15NO
Formula Weight 189.25
MDL Number MFCD01075691
Beilstein 4310531
Synonym 4'-(1-Pyrrolidino)acetophenone
Chemical Name or Material 4'-(1-Pyrrolidinyl)acetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1540806 SDS Alfa Aesar™
L1540806
5g Each for $84.22
AAL1540814 SDS Alfa Aesar™
L1540814
25g Each for $294.70
Specifications Description & Specifications

Chemical Identifiers

21557-09-5
189.25
4310531
4'-(1-Pyrrolidinyl)acetophenone
C12H15NO
MFCD01075691
4'-(1-Pyrrolidino)acetophenone

Specifications

5g
C12H15NO
MFCD01075691
4'-(1-Pyrrolidino)acetophenone
4'-(1-Pyrrolidinyl)acetophenone
21557-09-5
189.25
4310531
128° to 130°C
≥98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning