missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Piperidinylmethyl)benzeneboronic acid pinacol ester

$87.20 - $333.59

Chemical Identifiers

CAS 859833-22-0
Molecular Formula C18H28BNO2
Formula Weight 301.23
MDL Number MFCD08271932
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
Chemical Name or Material 4-(1-Piperidinylmethyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5111403 SDS Alfa Aesar™
H5111403
1g Each for $87.20
AAH5111406 SDS Alfa Aesar™
H5111406
5g Each for $333.59
Specifications Description & Specifications

Chemical Identifiers

859833-22-0
301.23
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
C18H28BNO2
MFCD08271932
4-(1-Piperidinylmethyl)benzeneboronic acid pinacol ester

Specifications

1g
C18H28BNO2
MFCD08271932
4-(1-Piperidinylmethyl)benzeneboronic acid pinacol ester
859833-22-0
301.23
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine