missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester

$45.41 - $704.99

Chemical Identifiers

CAS 938043-31-3
Molecular Formula C18H26BNO3
Formula Weight 315.21
MDL Number MFCD09027083
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine
Chemical Name or Material 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5111103 SDS Alfa Aesar™
H5111103
1g Each for $45.41
AAH5111106 SDS Alfa Aesar™
H5111106
5g Each for $173.93
AAH5111114 SDS Alfa Aesar™
H5111114
25g Each for $704.99
Specifications Description & Specifications

Chemical Identifiers

938043-31-3
315.21
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine
C18H26BNO3
MFCD09027083
4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester

Specifications

1g
C18H26BNO3
MFCD09027083
4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester
938043-31-3
315.21
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine