Learn More
4-(1-Piperidinyl)benzonitrile, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6120603
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-(1-Piperidinyl)benzonitrile | |
White | |
MFCD02575368 | |
UN3439 | |
ZEPXHFFGXQFUDP-UHFFFAOYSA-N | |
4-piperidin-1-ylbenzonitrile | |
766771 | |
98% |
1204-85-9 | |
C12H14N2 | |
1 g | |
4-piperidin-1-yl benzonitrile, 4-1-piperidinyl benzonitrile, 4-piperidin-1-yl-benzonitrile, 4-piperidinobenzonitrile, benzonitrile,4-1-piperidinyl, 4-piperidylbenzenecarbonitrile, acmc-209a7n, 4-piperidin-1-ylbenzenecarbonitrile | |
C1CCN(CC1)C2=CC=C(C=C2)C#N | |
186.258 | |
186.26 | |
Solid |
Chemical Identifiers
1204-85-9 | |
186.258 | |
ZEPXHFFGXQFUDP-UHFFFAOYSA-N | |
766771 | |
C1CCN(CC1)C2=CC=C(C=C2)C#N |
C12H14N2 | |
MFCD02575368 | |
4-piperidin-1-yl benzonitrile, 4-1-piperidinyl benzonitrile, 4-piperidin-1-yl-benzonitrile, 4-piperidinobenzonitrile, benzonitrile,4-1-piperidinyl, 4-piperidylbenzenecarbonitrile, acmc-209a7n, 4-piperidin-1-ylbenzenecarbonitrile | |
4-piperidin-1-ylbenzonitrile |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures