Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Piperidinyl)aniline, 97+%

Chemical Identifiers

Chemical Name or Material 4-(1-Piperidinyl)aniline
CAS 2359-60-6
Molecular Formula C11H16N2
Formula Weight 176.26
MDL Number MFCD00051688
UN Number UN2811
Beilstein 139525
Synonym 1-(4-Aminophenyl)piperidine; 4-(1-Piperidino)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0676703

  SDS
alfa aesar™
L0676703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0676706

  SDS
alfa aesar™
L0676706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Chemical Name or Material 4-(1-Piperidinyl)aniline
Assay ≥97%
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Danger
CAS 2359-60-6
MDL Number MFCD00051688
Boiling Point 140° to 142°C (1mmHg)
UN Number UN2811
Quantity 1g
Melting Point 26° to 28°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Molecular Formula C11H16N2
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 139525
Synonym 1-(4-Aminophenyl)piperidine; 4-(1-Piperidino)aniline
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.