missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Piperazinyl)aniline, 97%

Manufacturer:  Alfa Aesar™ H55224MD

 View more versions of this product

Catalog No. AAH55224MD



Description

Description

Applications
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Reactive with oxidizing agents. Keep at room temperature.
Specifications

Specifications

4-(1-Piperazinyl)aniline
119°C to 123°C
67455-41-8
MFCD01365906
VAVOYRCCWLRTMS-UHFFFAOYSA-N
4-piperazin-1-ylaniline
422925
Powder
Purple to White
250mg
C10H15N3
1-4-aminophenyl piperazine
C1CN(CCN1)C2=CC=C(C=C2)N
177.251
177.25
97%

Chemical Identifiers

67455-41-8
177.251
VAVOYRCCWLRTMS-UHFFFAOYSA-N
422925
C1CN(CCN1)C2=CC=C(C=C2)N
C10H15N3
MFCD01365906
1-4-aminophenyl piperazine
4-piperazin-1-ylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents